Learn More
VAISALA INC HH Humidity/Temperature Meter w/Remote Short Probe & Handle
Catalog No. NC1097679
Encompass_Preferred
The HM40 is an ideal everyday spot-checking tool for HVAC professionals. The proven Vaisala HUMICAP 180R sensor provides reliable measurement while the user interface with graphical trend display makes daily operation quick and easy.
Catalog No. NC1097679 Supplier VAISALA INC Supplier No. HM40B1AB
The HM40 is an ideal everyday spot-checking tool for HVAC professionals. The proven Vaisala HUMICAP 180R sensor provides reliable measurement while the user interface with graphical trend display makes daily operation quick and easy.