Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Val-Ala-PAB-MMAE | 1912408-92-4 | 98.9% | 1,037.33 g/mol | C55H88N8O11 | 10 MG
SDP

Catalog No. 5000204410
Encompass_Preferred

Val-Ala-PAB-MMAE is a drug-linker conjugate that couples the cleavable Val-Ala-PAB peptide linker to monomethyl auristatin E (MMAE), a potent tubulin inhibitor, and is supplied for antibody-drug conjugate research and development.

  • Drug-linker conjugate of Val-Ala-PAB and MMAE.
  • Purity 98.9%.
  • Molecular weight 1,037.33 g/mol; formula C55H88N8O11.
  • Supplied in multiple pack sizes, including 10 mg.
  • Intended for research use in antibody-drug conjugate development.

Catalog No. 50-002-04410 Supplier Medchemexpress LLC Supplier No. HY15303210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Val-Ala-PAB-MMAE is a drug-linker conjugate that couples the cleavable Val-Ala-PAB peptide linker to monomethyl auristatin E (MMAE), a potent tubulin inhibitor, and is supplied for antibody-drug conjugate research and development.

  • Drug-linker conjugate of Val-Ala-PAB and MMAE.
  • Purity 98.9%.
  • Molecular weight 1,037.33 g/mol; formula C55H88N8O11.
  • Supplied in multiple pack sizes, including 10 mg.
  • Intended for research use in antibody-drug conjugate development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.