Learn More
Medchemexpress LLC Val-Ala-PABC-Exatecan (trifluoroacetate) | 2928571-45-1 | 868.83 g·mol⁻¹ | C42H44F4N6O10 | 5 MG
SDP
Catalog No. 5000202121
Encompass_Preferred
Val-Ala-PABC-Exatecan (trifluoroacetate) is a drug-linker conjugate that combines a cleavable Val-Ala-PABC peptide linker with the topoisomerase I inhibitor exatecan. Supplied as the trifluoroacetate salt in milligram quantities, it is intended for research use in antibody-drug conjugate (ADC) synthesis and conjugation workflows.
  • Contains a cleavable Val-Ala-PABC peptide linker.
  • Delivers exatecan, a topoisomerase I inhibitor payload.
  • Supplied as the trifluoroacetate salt for handling stability.
  • Available in milligram quantities suitable for conjugation workflows.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-02121 Supplier Medchemexpress LLC Supplier No. HY147095A5MG
Val-Ala-PABC-Exatecan (trifluoroacetate) is a drug-linker conjugate that combines a cleavable Val-Ala-PABC peptide linker with the topoisomerase I inhibitor exatecan. Supplied as the trifluoroacetate salt in milligram quantities, it is intended for research use in antibody-drug conjugate (ADC) synthesis and conjugation workflows.
  • Contains a cleavable Val-Ala-PABC peptide linker.
  • Delivers exatecan, a topoisomerase I inhibitor payload.
  • Supplied as the trifluoroacetate salt for handling stability.
  • Available in milligram quantities suitable for conjugation workflows.
  • Intended for research use only; not for human or clinical use.