Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Val-cit-amide-ph-maytansine | 98.0% | 1055.61 | C51H71ClN8O14 | 50 MG
SDP

Catalog No. 5000204367
Encompass_Preferred

Val-Cit-amide-Ph-Maytansine is a maytansine-derived payload bearing a valine-citrulline cleavable linker amide, supplied as a white to off-white solid for antibody-drug conjugate research and conjugation workflows.

  • Cleavable valine-citrulline linker amide suitable for ADC conjugation
  • Maytansine-derived cytotoxic payload with high potency
  • Purity approximately 98.0%
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation may be required
  • Available in multiple pack sizes, including 50 mg
  • Stable as powder at -20°C for long-term storage

Catalog No. 50-002-04367 Supplier Medchemexpress LLC Supplier No. HY15689750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Val-Cit-amide-Ph-Maytansine is a maytansine-derived payload bearing a valine-citrulline cleavable linker amide, supplied as a white to off-white solid for antibody-drug conjugate research and conjugation workflows.

  • Cleavable valine-citrulline linker amide suitable for ADC conjugation
  • Maytansine-derived cytotoxic payload with high potency
  • Purity approximately 98.0%
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation may be required
  • Available in multiple pack sizes, including 50 mg
  • Stable as powder at -20°C for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.