Learn More
Medchemexpress LLC Val-Cit-PAB-MMAF (sodium) | 863971-56-6 | 98.4% | 1159.39 | C58H91N10NaO13 | 50 MG
SDP
Catalog No. 5000202728
Encompass_Preferred
Val-Cit-PAB-MMAF sodium is an antibody-drug conjugate (ADC) drug-linker conjugate that links the protease-cleavable Val-Cit-PAB peptide linker to the tubulin inhibitor monomethyl auristatin F (MMAF). Supplied as the sodium salt, it is intended for use as an ADC building block for research applications, supporting in vitro formulation and in vivo dosing studies.
  • Protease-cleavable Val-Cit-PAB linker enabling cathepsin B-triggered payload release.
  • MMAF payload provides potent tubulin polymerization inhibition.
  • High purity suitable for research applications (98.4%).
  • Soluble in DMSO and formulatable for in vivo administration with appropriate vehicles.
  • Available in multiple small milligram package sizes for ADC synthesis and evaluation.
Catalog No. 50-002-02728 Supplier Medchemexpress LLC Supplier No. HY4417750MG
Val-Cit-PAB-MMAF sodium is an antibody-drug conjugate (ADC) drug-linker conjugate that links the protease-cleavable Val-Cit-PAB peptide linker to the tubulin inhibitor monomethyl auristatin F (MMAF). Supplied as the sodium salt, it is intended for use as an ADC building block for research applications, supporting in vitro formulation and in vivo dosing studies.
  • Protease-cleavable Val-Cit-PAB linker enabling cathepsin B-triggered payload release.
  • MMAF payload provides potent tubulin polymerization inhibition.
  • High purity suitable for research applications (98.4%).
  • Soluble in DMSO and formulatable for in vivo administration with appropriate vehicles.
  • Available in multiple small milligram package sizes for ADC synthesis and evaluation.