Learn More
Medchemexpress LLC Val-cit-pab-mmaf (sodium) | 98.4% | 1159.39 g/mol | C58H91N10NaO13 | 5 MG
SDP
Catalog No. 5000202729
Encompass_Preferred
Val-Cit-PAB-MMAF sodium is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links the protease-cleavable peptide Val-Cit-PAB to the cytotoxic payload monomethyl auristatin F (MMAF). It is supplied as the sodium salt and intended for research use.
  • Protease-cleavable Val-Cit-PAB linker coupled to MMAF.
  • Intended for ADC research and conjugation workflows.
  • High purity suitable for research applications.
  • Soluble in DMSO at high concentration; in vivo formulations achieve ≥5 mg/mL with solvent mixtures.
  • Provided in small research pack sizes for preclinical experiments.
Catalog No. 50-002-02729 Supplier Medchemexpress LLC Supplier No. HY441775MG
Val-Cit-PAB-MMAF sodium is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links the protease-cleavable peptide Val-Cit-PAB to the cytotoxic payload monomethyl auristatin F (MMAF). It is supplied as the sodium salt and intended for research use.
  • Protease-cleavable Val-Cit-PAB linker coupled to MMAF.
  • Intended for ADC research and conjugation workflows.
  • High purity suitable for research applications.
  • Soluble in DMSO at high concentration; in vivo formulations achieve ≥5 mg/mL with solvent mixtures.
  • Provided in small research pack sizes for preclinical experiments.