Learn More
Cayman Chemical Val-CIt-PAB-OH 100mg
SDP
Catalog No. 502910467
Encompass_Preferred
A peptide linker molecule; cleaved by cathepsin B; has been used in the synthesis of ADCs containing KGP05 or MMAD, as well as α-galactosylceramide or folic acid conjugates that target peptide antigens to natural killer T cells or CRM1 inhibitors to cancer cells, respectively
Catalog No. 50-291-0467 Supplier Cayman Chemical Supplier No. 34818100 MG
A peptide linker molecule; cleaved by cathepsin B; has been used in the synthesis of ADCs containing KGP05 or MMAD, as well as α-galactosylceramide or folic acid conjugates that target peptide antigens to natural killer T cells or CRM1 inhibitors to cancer cells, respectively