Learn More
Medchemexpress LLC Val-cit-PAB-OH | 159857-79-1 | 379.45 | C18H29N5O4 | 25 G
SDP
Catalog No. 5000201895
Encompass_Preferred
Val-cit-PAB-OH is a cleavable, protease-sensitive antibody-drug conjugate (ADC) linker that couples cytotoxic payloads to antibodies through a valine-citrulline dipeptide and a para-aminobenzyl (PAB) self-immolative spacer. It is designed to remain stable in circulation and release payloads upon intracellular protease cleavage, supporting targeted delivery in ADC synthesis.
  • Cleavable by intracellular proteases via valine-citrulline mechanism.
  • Stable in systemic circulation, reducing premature payload release.
  • Enables self-immolative payload release through a PAB spacer.
  • Compatible with common conjugation chemistries for ADC construction.
  • Available in gram-scale quantities for research and scale-up synthesis.
  • Supplied with manufacturer storage guidance for long-term stability.
Catalog No. 50-002-01895 Supplier Medchemexpress LLC Supplier No. HY1236225G
Val-cit-PAB-OH is a cleavable, protease-sensitive antibody-drug conjugate (ADC) linker that couples cytotoxic payloads to antibodies through a valine-citrulline dipeptide and a para-aminobenzyl (PAB) self-immolative spacer. It is designed to remain stable in circulation and release payloads upon intracellular protease cleavage, supporting targeted delivery in ADC synthesis.
  • Cleavable by intracellular proteases via valine-citrulline mechanism.
  • Stable in systemic circulation, reducing premature payload release.
  • Enables self-immolative payload release through a PAB spacer.
  • Compatible with common conjugation chemistries for ADC construction.
  • Available in gram-scale quantities for research and scale-up synthesis.
  • Supplied with manufacturer storage guidance for long-term stability.