Learn More
Medchemexpress LLC Val-cit-PAB-OH | 159857-79-1 | MFCD22381209 | 99.6% | 379.45 g/mol | C18H29N5O4 | 100 MG
SDP
Catalog No. 5000203735
Encompass_Preferred
Val-cit-PAB-OH is a protease-cleavable, degradable antibody-drug conjugate (ADC) linker (valine-citrulline-p-aminobenzyl alcohol) supplied as a white to off-white solid for research use. It is designed for incorporation into ADCs to enable payload release following enzymatic cleavage and is provided with supporting documentation including a datasheet, certificate of analysis, and safety data sheet.
  • Degradable protease-cleavable linker enabling controlled payload release.
  • Suitable for ADC synthesis and linker-drug conjugation workflows.
  • High purity appropriate for research applications.
  • Soluble in DMSO at ≥ 100 mg/mL for preparation of concentrated stocks.
  • Stable when stored at 4°C under inert atmosphere; in solution, store at -80°C for long-term storage.
  • Supplied as a white to off-white solid for handling and formulation flexibility.
Catalog No. 50-002-03735 Supplier Medchemexpress LLC Supplier No. HY12362100MG
Val-cit-PAB-OH is a protease-cleavable, degradable antibody-drug conjugate (ADC) linker (valine-citrulline-p-aminobenzyl alcohol) supplied as a white to off-white solid for research use. It is designed for incorporation into ADCs to enable payload release following enzymatic cleavage and is provided with supporting documentation including a datasheet, certificate of analysis, and safety data sheet.
  • Degradable protease-cleavable linker enabling controlled payload release.
  • Suitable for ADC synthesis and linker-drug conjugation workflows.
  • High purity appropriate for research applications.
  • Soluble in DMSO at ≥ 100 mg/mL for preparation of concentrated stocks.
  • Stable when stored at 4°C under inert atmosphere; in solution, store at -80°C for long-term storage.
  • Supplied as a white to off-white solid for handling and formulation flexibility.