Learn More
Cayman Chemical Val-Cit-PAB-OH, Purity: ≥98%, CAS: 159857-79-1, 50 mg
SDP
Catalog No. 502911061
Encompass_Preferred
A peptide linker molecule; cleaved by cathepsin B; has been used in the synthesis of ADCs containing KGP05 or MMAD, as well as α-galactosylceramide or folic acid conjugates that target peptide antigens to natural killer T cells or CRM1 inhibitors to cancer cells, respectively
Catalog No. 50-291-1061 Supplier Cayman Chemical Supplier No. 3481850 MG
A peptide linker molecule; cleaved by cathepsin B; has been used in the synthesis of ADCs containing KGP05 or MMAD, as well as α-galactosylceramide or folic acid conjugates that target peptide antigens to natural killer T cells or CRM1 inhibitors to cancer cells, respectively