Learn More
Bachem (Val5)-Angiotensin II
SDP
Supplier:  Bachem 4008960.0025
Encompass_Preferred
(Val5)-Angiotensin II, H-Asp-Arg-Val-Tyr-Val-His-Pro-Phe-OH, 58-49-1, C49H69N13O12, Mr 1032.17, 25mg. [Val5] Angiotensin II, human.
Catalog No. 50-259-533