Learn More
CPC Scientific H-Asp-Arg-Val-Tyr-Ile-His-Pro-Val-His-Leu-Leu-Tyr-Tyr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110365
Encompass
ONE-LETTER SEQUENCE: DRVYIHPVHLLYYSMOLECULAR FORMULA: C85H123N21O21MOLECULAR WEIGHT:1775.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-211-0365 Supplier CPC Scientific Supplier No. RENI009A
ONE-LETTER SEQUENCE: DRVYIHPVHLLYYSMOLECULAR FORMULA: C85H123N21O21MOLECULAR WEIGHT:1775.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular