Learn More
Selleck Chemical LLC Valaciclovir HCl S1876-50mg
SDP
Catalog No. 503109356
Encompass_Preferred
Valaciclovir HCl an aciclovir prodrug inhibits activity of virus DNA polymerase used to treat infections caused by herpes simplex virus (HSV) and varicella zoster virus and for prophylaxis against cytomegalovirus (CMV)
Catalog No. 50-310-9356 Supplier Selleck Chemical LLC Supplier No. S187650MG
Add to Cart
Add to Cart
Valaciclovir HCl an aciclovir prodrug inhibits activity of virus DNA polymerase used to treat infections caused by herpes simplex virus (HSV) and varicella zoster virus and for prophylaxis against cytomegalovirus (CMV)