Learn More
Medchemexpress LLC Valacyclovir-d8 hydrochloride | 1279033-32-7 | 98.0% | 368.85 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17425AS1MG
Encompass_Preferred
Valacyclovir-d8 hydrochloride is a deuterium-labeled, orally active antiviral agent. It is effective against herpes simplex, herpes zoster, and herpes B, and functions as a proagent of Aciclovir. Intended for research use only, this compound serves as a tracer or internal standard for quantitative analysis, utilizing stable heavy isotopes to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Orally active antiviral agent
  • Effective against herpes simplex, herpes zoster, and herpes B
  • Inhibits HSV-1 W
  • Proagent of Aciclovir
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-83969