Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valacyclovir hydrochloride | 124832-27-5 | 98.7% | 360.80 | 1 ML
SDP

Catalog No. 5000386101
Encompass_Preferred

Valacyclovir hydrochloride is an orally active antiviral agent effective against herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 with an IC50 of 2.9 μg/mL and is a proagent of Aciclovir.

  • Orally active antiviral agent
  • Effective against herpes simplex, herpes zoster, and herpes B
  • Inhibits HSV-1 with an IC50 of 2.9 μg/mL
  • Proagent of Aciclovir
  • Soluble in water and DMSO
  • Appearance is a white to off-white solid

Catalog No. 50-003-86101 Supplier Medchemexpress LLC Supplier No. HY17425A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valacyclovir hydrochloride is an orally active antiviral agent effective against herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 with an IC50 of 2.9 μg/mL and is a proagent of Aciclovir.

  • Orally active antiviral agent
  • Effective against herpes simplex, herpes zoster, and herpes B
  • Inhibits HSV-1 with an IC50 of 2.9 μg/mL
  • Proagent of Aciclovir
  • Soluble in water and DMSO
  • Appearance is a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.