Learn More
Medchemexpress LLC Valacyclovir hydrochloride | 124832-27-5 | 98.7% | 360.80 | 50 MG
SDP
Catalog No. 5000385302
Encompass_Preferred
Valacyclovir hydrochloride is an orally active antiviral agent effective against herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 with an IC50 of 2.9 μg/mL and is a prodrug of Aciclovir.
  • Administered once a day
  • Effective for treating recurrences of genital herpes
  • 1 g daily dose as effective as 2 g daily for recurrences
Catalog No. 50-003-85302 Supplier Medchemexpress LLC Supplier No. HY17425A50MG
Valacyclovir hydrochloride is an orally active antiviral agent effective against herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 with an IC50 of 2.9 μg/mL and is a prodrug of Aciclovir.
  • Administered once a day
  • Effective for treating recurrences of genital herpes
  • 1 g daily dose as effective as 2 g daily for recurrences