Learn More
Medchemexpress LLC Valacyclovir hydrochloride | 124832-27-5 | 99.9% | 360.80 | 1 G
SDP
Supplier:  Medchemexpress LLC HY17425A1G
Encompass_Preferred
Valacyclovir hydrochloride (Valaciclovir hydrochloride) is an orally active antiviral agent effective against herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 W with a concentration of 50=2.9 μg/ml. Valacyclovir hydrochloride acts as a proagent of Aciclovir.
  • Orally active antiviral agent
  • Effective against herpes simplex, herpes zoster, and herpes B
  • Inhibits HSV-1 W
  • Proagent of Aciclovir
Catalog No. 50-003-85970