Learn More
Medchemexpress LLC Valacyclovir hydrochloride | 124832-27-5 | 99.9% | 360.80 | 100 MG
SDP
Catalog No. 5000386102
Encompass_Preferred
Valacyclovir hydrochloride is an orally active antiviral agent for herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 W (50=2.9 μg/ml) and is a proagent of Aciclovir.
  • IC50 and target: HSV-1, 2.9 μg/mL (IC50).
  • In vitro: Uptake is concentration dependent and saturable, with hPEPT1 dominating intestinal transport.
  • In vivo: Effective for treating genital herpes (1 g twice daily) and recurrences. A 1 g daily dose is as effective as 2 g daily and can be administered once a day. Studies in immunosuppressed mice showed reduced erythema, eliminated ear paralysis, ear lesions, and death.
Catalog No. 50-003-86102 Supplier Medchemexpress LLC Supplier No. HY17425A100MG
Valacyclovir hydrochloride is an orally active antiviral agent for herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 W (50=2.9 μg/ml) and is a proagent of Aciclovir.
  • IC50 and target: HSV-1, 2.9 μg/mL (IC50).
  • In vitro: Uptake is concentration dependent and saturable, with hPEPT1 dominating intestinal transport.
  • In vivo: Effective for treating genital herpes (1 g twice daily) and recurrences. A 1 g daily dose is as effective as 2 g daily and can be administered once a day. Studies in immunosuppressed mice showed reduced erythema, eliminated ear paralysis, ear lesions, and death.