Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valacyclovir hydrochloride | 124832-27-5 | MFCD01861507 | 99.0% | 10 MG
SDP

Catalog No. 5000297952
Encompass_Preferred

Valacyclovir hydrochloride is the hydrochloride salt of valacyclovir, an orally active antiviral prodrug of acyclovir used against herpes simplex and herpes zoster. Supplied for research and analytical use, this solid reagent is intended as a reference standard or for antiviral studies and method development.

  • Orally active antiviral prodrug of acyclovir
  • Hydrochloride salt supplied as a solid for research use
  • Analytical standard with reported purity 99.0% (HPLC)
  • Molecular weight 360.80 and formula C13H21ClN6O4
  • Suitable for antiviral research and reference standard applications
  • Available in small pack sizes for analytical work

Catalog No. 50-002-97952 Supplier Medchemexpress LLC Supplier No. HY17425AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valacyclovir hydrochloride is the hydrochloride salt of valacyclovir, an orally active antiviral prodrug of acyclovir used against herpes simplex and herpes zoster. Supplied for research and analytical use, this solid reagent is intended as a reference standard or for antiviral studies and method development.

  • Orally active antiviral prodrug of acyclovir
  • Hydrochloride salt supplied as a solid for research use
  • Analytical standard with reported purity 99.0% (HPLC)
  • Molecular weight 360.80 and formula C13H21ClN6O4
  • Suitable for antiviral research and reference standard applications
  • Available in small pack sizes for analytical work

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.