Learn More
Medchemexpress LLC Valategrast | 220847-86-9 | MFCD18782681 | 98.6% | C30H32Cl3N3O4 | 10MG
SDP
Catalog No. 5000259835
Encompass_Preferred
Valategrast (R-411 free base) is a potent, orally active dual antagonist of integrins α4β1 (VLA-4) and α4β7 provided for research use in inflammation and respiratory disease studies. It is supplied as a solid, characterized material for preclinical and in vitro investigations; not for human therapeutic use.
  • Potent orally active dual integrin antagonist useful for mechanistic studies.
  • Applicable to COPD and asthma research models.
  • High purity solid suitable for analytical and biological assays.
  • Available in multiple small pack sizes for research flexibility.
  • Characterized with molecular weight, formula, and purity data for reproducibility.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-59835 Supplier Medchemexpress LLC Supplier No. HY1419010MG
Valategrast (R-411 free base) is a potent, orally active dual antagonist of integrins α4β1 (VLA-4) and α4β7 provided for research use in inflammation and respiratory disease studies. It is supplied as a solid, characterized material for preclinical and in vitro investigations; not for human therapeutic use.
  • Potent orally active dual integrin antagonist useful for mechanistic studies.
  • Applicable to COPD and asthma research models.
  • High purity solid suitable for analytical and biological assays.
  • Available in multiple small pack sizes for research flexibility.
  • Characterized with molecular weight, formula, and purity data for reproducibility.
  • For research use only; not for human therapeutic use.