Learn More
Medchemexpress LLC Valbenazine | 1025504-45-3 | 98.6% | 418.57 | 200 MG
SDP
Catalog No. 5000390758
Encompass_Preferred
Valbenazine is a vesicular monoamine transporter 2 (VMAT2) inhibitor with a Ki of 110-190 nM. It exhibits VMAT2 binding affinity in rat striatum and human platelets. In vivo, Valbenazine induces ptosis and increases plasma prolactin in rats.
  • VMAT2 inhibitor.
  • Exhibits VMAT2 binding affinity in rat striatum and human platelets.
  • Induces ptosis and increases plasma prolactin in rats.
  • Available in solid and solution forms.
  • Can be stored long-term as powder at -20°C (3 years) or 4°C (2 years).
  • In solvent, stock solutions can be stored at -80°C (2 years) or -20°C (1 year).
Catalog No. 50-003-90758 Supplier Medchemexpress LLC Supplier No. HY16771200MG
Valbenazine is a vesicular monoamine transporter 2 (VMAT2) inhibitor with a Ki of 110-190 nM. It exhibits VMAT2 binding affinity in rat striatum and human platelets. In vivo, Valbenazine induces ptosis and increases plasma prolactin in rats.
  • VMAT2 inhibitor.
  • Exhibits VMAT2 binding affinity in rat striatum and human platelets.
  • Induces ptosis and increases plasma prolactin in rats.
  • Available in solid and solution forms.
  • Can be stored long-term as powder at -20°C (3 years) or 4°C (2 years).
  • In solvent, stock solutions can be stored at -80°C (2 years) or -20°C (1 year).