Learn More
Cayman Chemical Valbenazine, Purity: ≥98%, CAS: 1025504-45-3, 50 mg
SDP
Catalog No. 502935229
Encompass_Preferred
An inhibitor of VMAT2 (Kis = 110 and 150 nM in rat striatum homogenates and human platelets, respectively); selective for VMAT2 over a panel of 80 receptors, transporters, and ion channels; increases ptosis in rats
Catalog No. 50-293-5229 Supplier Cayman Chemical Supplier No. 2680950 MG
An inhibitor of VMAT2 (Kis = 110 and 150 nM in rat striatum homogenates and human platelets, respectively); selective for VMAT2 over a panel of 80 receptors, transporters, and ion channels; increases ptosis in rats