Learn More
Thermo Scientific™ Valco™ Injector Model C6W
Catalog No. 03170308 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Injector/Switching Valve
Rotor Seal Valve
Volume (Metric) Sample:
100 μL
N/A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Volume (Metric) Sample
03-170-308 Rotor Seal Valve N/A
03-052-441 Injector/Switching Valve 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-170-308 Supplier Thermo Scientific™ Supplier No. SSAC6W
Add to Cart
Add to Cart
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve.
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve. Available with manual or a microelectric actuator. Has 1/16 in. fittings and six 0.010 inch ports.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) Sample N/A
Description Replacement Rotor
Type Rotor Seal Valve
No. of Ports 6 Ports
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.