Learn More
MilliporeSigma™ Supelco™ Valco™ Precision Sampling Plunger Tips
Catalog No. 111011715 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Size (Metric):
1 mL
Catalog No. Size (Metric)
11-101-1715 1 mL
1 options
Catalog No. 11-101-1715 Supplier MilliporeSigma™ Supelco™ Supplier No. 21951
Add to Cart
Add to Cart
For series A, A-2, C, and D Pressure-Lok Gas syringes. For 25 μL to 250 μL syringes, factory repair only.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Size (Metric) 1 mL
Description Plunger tip for 1 mL syringe
For Use With (Equipment) Series A, A-2, C, and D Precision Sampling Syringe and VICI Series A, A-2, C, and D Pressure-Lok Gas Syringes
Valco is a registered trademark of Valco Instruments Co. Inc.