Learn More
Perkin Elmer US LLC Valco Stainless Steel Male Nut, 0.0625 in
Catalog No. 502112494
Encompass_Preferred
Cannot be used with fused silica adapters (N9302846 and N9302847). This nut is included in the Arnel Services Spares Kit (N6100726). Hex: 1/4 inch
Catalog No. 50-211-2494 Supplier Perkin Elmer US LLC Supplier No. N9302832
Cannot be used with fused silica adapters (N9302846 and N9302847). This nut is included in the Arnel Services Spares Kit (N6100726). Hex: 1/4 inch