Learn More
Trajan Scientific VALCO VALVE 20000
Catalog No. NC1943598
Encompass
VALCO Cheminert 2 position “Injection” Valve, with 6 ports; 0.25 mm ports, Max pressure: 20K psi, Valcon E5 – Suggested Rotor: VAL.C72-15R6 Suggested Stator: VAL.C72V-1C76
Catalog No. NC1943598 Supplier Trajan Scientific Supplier No. VAL.C72VU1576DCTC
VALCO Cheminert 2 position “Injection” Valve, with 6 ports; 0.25 mm ports, Max pressure: 20K psi, Valcon E5 – Suggested Rotor: VAL.C72-15R6 Suggested Stator: VAL.C72V-1C76