Learn More
Med Vet International Valcor (Doramectin And Levamisole) Injection, 250mL
Catalog No. 502828170
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
VALCOR™ (doramectin and levamisole injection) is for the treatment and control of gastrointestinal roundworms, lungworms, eyeworms, grubs, sucking lice and mange mites in beef cattle 2 months of age and older and in replacement dairy heifers less than 20 months of age. Not for use in beef bulls intended for breeding over 1 year of age, dairy calves and veal calvesParasiticide for subcutaneous injection in cattle
Catalog No. 50-282-8170 Supplier Med Vet International Supplier No. RXVALCOR250ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
VALCOR™ (doramectin and levamisole injection) is for the treatment and control of gastrointestinal roundworms, lungworms, eyeworms, grubs, sucking lice and mange mites in beef cattle 2 months of age and older and in replacement dairy heifers less than 20 months of age. Not for use in beef bulls intended for breeding over 1 year of age, dairy calves and veal calvesParasiticide for subcutaneous injection in cattle