Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Valdecoxib

Catalog No. AC462522500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462522500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462522500 Supplier Thermo Scientific Chemicals Supplier No. 462522500

missing translation for 'validMainframeMsg'

CAS: 181695-72-7 | C16H14N2O3S | 314.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181695-72-7
Molecular Formula C16H14N2O3S
Molecular Weight (g/mol) 314.46

Specifications

Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in acetone, ethanol & methanol
Formula Weight 314.46
Chemical Name or Material Valdecoxib
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of damaging the unborn child.

May cause damage to organs through prolonged or repeated exposure.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.