Learn More
Medchemexpress LLC HY-13598 5mg Medchemexpress, Valecobulin CAS:1188371-47-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135985MG
Encompass_Preferred
Medchemexpress, HY-13598 5mg Valecobulin CAS:1188371-47-2 Valecobulin (CKD516) is a valine prodrug of (S516) and a vascular disrupting agent (VDA). Valecobulin is a potent β-tubulin polymerization inhibitor with marked antitumor activity against murine and human solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9884