Learn More
Medchemexpress LLC HY-13598A 5mg Medchemexpress, Valecobulin (hydrochloride) CAS:1240321-53-2 Purity:>98%
SDP
Catalog No. 501969885
Encompass_Preferred
Medchemexpress, HY-13598A 5mg Valecobulin (HCl) CAS:1240321-53-2 Valecobulin hydrochloride (CKD-516 hydrochloride) is a valine prodrug of S516 (HY-130233) and a vascular disrupting agent (VDA). Valecobulin hydrochloride is a potent β-tubulin polymerization inhibitor with marked antitumor activity against murine and human solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9885 Supplier Medchemexpress LLC Supplier No. HY13598A5MG
Medchemexpress, HY-13598A 5mg Valecobulin (HCl) CAS:1240321-53-2 Valecobulin hydrochloride (CKD-516 hydrochloride) is a valine prodrug of S516 (HY-130233) and a vascular disrupting agent (VDA). Valecobulin hydrochloride is a potent β-tubulin polymerization inhibitor with marked antitumor activity against murine and human solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.