Learn More
Medchemexpress LLC Valecobulin hydrochloride | 1240321-53-2 | MFCD19311027 | 98.5% | 573.06 | 50 MG
SDP
Catalog No. 5000364478
Encompass_Preferred
Valecobulin hydrochloride (CKD-516 hydrochloride) is a valine proagent of S516 (HY-130233) and a vascular disrupting agent (VDA). It is a potent β-tubulin polymerization inhibitor with marked antitumor activity against murine and human solid tumors.
  • Valine proagent of S516
  • Vascular disrupting agent (VDA)
  • Potent β-tubulin polymerization inhibitor
  • Marked antitumor activity against murine and human solid tumors
Catalog No. 50-003-64478 Supplier Medchemexpress LLC Supplier No. 50MGHY13598A
Valecobulin hydrochloride (CKD-516 hydrochloride) is a valine proagent of S516 (HY-130233) and a vascular disrupting agent (VDA). It is a potent β-tubulin polymerization inhibitor with marked antitumor activity against murine and human solid tumors.
  • Valine proagent of S516
  • Vascular disrupting agent (VDA)
  • Potent β-tubulin polymerization inhibitor
  • Marked antitumor activity against murine and human solid tumors