Learn More
Medchemexpress LLC Valecobulin hydrochloride | 1240321-53-2 | MFCD19311027 | 98.5% | C26H29ClN6O5S | 10MG
SDP
Catalog No. 5000260280
Encompass_Preferred
Valecobulin hydrochloride is a valine prodrug of S516 and a vascular disrupting agent that inhibits β-tubulin polymerization. It has reported antitumor activity against murine and human solid tumors and is supplied for laboratory research use in both solid and solution forms.
  • Potent β-tubulin polymerization inhibitor with antitumor activity.
  • Valine prodrug of S516, suitable for tubulin-targeted studies.
  • Vascular disrupting agent activity relevant to tumor biology research.
  • High purity (≈98.5%), appropriate for in vitro and in vivo experiments.
  • Available in small research pack sizes and solution formats.
  • Stable when sealed; long-term storage recommended at -80°C in solvent.
Catalog No. 50-002-60280 Supplier Medchemexpress LLC Supplier No. HY13598A10MG
Valecobulin hydrochloride is a valine prodrug of S516 and a vascular disrupting agent that inhibits β-tubulin polymerization. It has reported antitumor activity against murine and human solid tumors and is supplied for laboratory research use in both solid and solution forms.
  • Potent β-tubulin polymerization inhibitor with antitumor activity.
  • Valine prodrug of S516, suitable for tubulin-targeted studies.
  • Vascular disrupting agent activity relevant to tumor biology research.
  • High purity (≈98.5%), appropriate for in vitro and in vivo experiments.
  • Available in small research pack sizes and solution formats.
  • Stable when sealed; long-term storage recommended at -80°C in solvent.