Learn More
Julabo USA Inc VALEGRO 500 Chiller R290 500W
SDP
Catalog No. 5000077884
Encompass_Preferred
Chiller JULABO VALEGRO 500 115V/60Hz R290 Refrigerant -20 40 C Cooling 0 5kW at 20C 0 37kW at 0C 0 25kW at -10C 0 1kW at -20C Adjustable pump to 19Lpm 12 3PSI OLED touchscreen UI with USB-C RS232 and optional Ethernet USB data-logging integrated timer M16x1 pump fittings with 8 and 12mm barbs 1 5-3L bath tank 9 1x 15 7 x 17 3 in 57 3lbs
Catalog No. 50-000-77884 Supplier Julabo USA Inc Supplier No. 9610050.3
Add to Cart
Add to Cart
Request a Quote
Chiller JULABO VALEGRO 500 115V/60Hz R290 Refrigerant -20 40 C Cooling 0 5kW at 20C 0 37kW at 0C 0 25kW at -10C 0 1kW at -20C Adjustable pump to 19Lpm 12 3PSI OLED touchscreen UI with USB-C RS232 and optional Ethernet USB data-logging integrated timer M16x1 pump fittings with 8 and 12mm barbs 1 5-3L bath tank 9 1x 15 7 x 17 3 in 57 3lbs