Learn More
Medchemexpress LLC HY-109108A 5mg Medchemexpress, Valemetostat (tosylate) CAS:1809336-93-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109108A5MG
Encompass_Preferred
Medchemexpress, HY-109108A 5mg Valemetostat (tosylate) CAS:1809336-93-3 Valemetostat tosylate (DS-3201 tosylate) is a first-in-class EZH1/2 dual inhibitor, used in the research of relapsed/refractory peripheral T-cell lymphoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8683