Learn More
Valence Shell Electron Pair Repulsion Models
SCI_ED
Catalog No. S65371
Change view
Click to view available options
Vsepr2 molecular model
Catalog No.
S65371 Vsepr2 molecular model
1 options
Catalog No. S65371 Supplier Ryler Enterprises Supplier No. VSP2
Valence Shell Electron Pair Repulsion (VSEPR) models illustrate molecular geometry, resonance, and molecule polarity.
VSEPR 1 model
Supports students' investigation of the octet rule to create models of molecules with linear, tetrahedral, pyramidal, bent, and trigonal planar shapes.
Model includes:
  • Two carbons
  • Four oxygens
  • Five hydrogens
  • Three halogens
  • Three nitrogens
  • Two sulfurs
  • Six short tubes
  • Six long tubes
VSEPR 2 model
Helps students understand exceptions to the octet rule. Students will create octahedral, square planar, trigonal, hipyramidal, trigonal planar, linear, and bent molecules.
Model includes:
  • 2-2 trigonal bipyramidal
  • Two octahedral
  • Five oxygens
  • Three hydrogens
  • Seven halogens
  • Two nitrogens
  • Eight short tubes
  • Eight long tubes
Specifications
For Use With (Application) Illustrates molecular geometry, resonance and molecule polarity
Includes Red 4-peg Oxygen atom centers, green 4-peg Halogen atom centers, purple 5-peg Phosphorous atom centers, Silver 6-peg sulfur (xenon) atom centers, 2 yellow 4-peg sulfur atom centers, 1.25 in. PVC, clear bonds (single bonds), 1.625 in. PVC, clear bonds (double bonds), background information booklet, lab procedure, student response form and answer key
Product Type Model, Valence Shell Electron Pair Repulsion