Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Valencene, ≥80% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014228 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4228 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4228 Supplier MilliporeSigma Supelco Supplier No. 0680850MG
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 7-3-4630
Density 0.92 g/mL (at 25°C)
Flash Point 100°C
Linear Formula C15H24
MDL Number MFCD00075884
Boiling Point 274°C
Synonym (3 R,4 AS,5 R)-4 A,5-Dimethyl-3-isopropenyl-1,2,3,4,4 A,5,6,7-octahydronaphthalene
Type Analytical Standard
Quantity 50 mg
Formula Weight 204.35
Percent Purity ≥80% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.