Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valencene (Tech.), 70%, Thermo Scientific™

Catalog No. AC240830250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC240830250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC240830250 Supplier Thermo Scientific Chemicals Supplier No. 240830250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4635-08-9
Molecular Formula C5H6BrN3
Molecular Weight (g/mol) 188.03
InChI Key CMFSWSZEHYPECE-UHFFFAOYSA-N
Synonym 5-bromo-3,4-diaminopyridine, pyridine, 3-bromo-4,5-diamino, 5-bromo-pyridine-3,4-diamine, 3-bromo-4,5-diaminopyridine, 3,4-pyridinediamine, 5-bromo, 5-bromo-3,4-pyridinediamine, pubchem23608, 3,4-diamino-5-bromopyridine, 3-bromo-4,5-pyridinediamine
PubChem CID 78369
IUPAC Name 5-bromopyridine-3,4-diamine
SMILES C1=C(C(=C(C=N1)Br)N)N

Specifications

Color Colorless to Yellow
Density 0.9355
Infrared Spectrum Authentic
Refractive Index 1.5000 to 1.5150 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 0.91 to 0.961 (25°/25°C)
Formula Weight 204.36
Percent Purity ≥65%
Grade Technical
Physical Form Liquid
Chemical Name or Material Valencene, tech., 70%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.