Learn More
MSC Valeo General Purpose Gloves, X-Large, Goatskin, Black, 1 Pair
Catalog No. 5000788379
Encompass
These General Purpose Gloves are designed for a variety of tasks, offering a comfortable and secure fit. They feature a goatskin primary material and a nylon mesh back for breathability, providing general protection for various applications.
  • Made with goatskin primary material
  • Features nylon mesh back for breathability
  • X-Large size (numeric size 10)
  • Unlined with hook & loop cuff style
  • Padded goat leather palm for enhanced protection
  • Machine washable for easy care
  • Unisex design for general protection
Catalog No. 50-007-88379 Supplier MSC Supplier No. 00250308
These General Purpose Gloves are designed for a variety of tasks, offering a comfortable and secure fit. They feature a goatskin primary material and a nylon mesh back for breathability, providing general protection for various applications.
  • Made with goatskin primary material
  • Features nylon mesh back for breathability
  • X-Large size (numeric size 10)
  • Unlined with hook & loop cuff style
  • Padded goat leather palm for enhanced protection
  • Machine washable for easy care
  • Unisex design for general protection