Learn More
MSC Valeo Gloves, V140, X-Large, Leather, Full Fingered, 1 Pair
Catalog No. 5000796830
Encompass
The V140 General Purpose Gloves are designed for a variety of tasks, offering an X-Large size and a black color. Made with synthetic leather and an unlined interior, these gloves feature a hook & loop cuff style for a secure fit.
  • Machine washable
  • Neoprene knuckle pad
  • Reinforced fingertips
  • Stitching for enhanced durability and protection
  • Full-fingered design
  • Seamless construction
  • Suitable for general protection
  • Double-layer synthetic leather palm
Catalog No. 50-007-96830 Supplier MSC Supplier No. 07431984
The V140 General Purpose Gloves are designed for a variety of tasks, offering an X-Large size and a black color. Made with synthetic leather and an unlined interior, these gloves feature a hook & loop cuff style for a secure fit.
  • Machine washable
  • Neoprene knuckle pad
  • Reinforced fingertips
  • Stitching for enhanced durability and protection
  • Full-fingered design
  • Seamless construction
  • Suitable for general protection
  • Double-layer synthetic leather palm