Learn More
Valeo Fingerless Material Handling Cowhide Leather Gloves with Mesh Back
Catalog No. 19048692C
Encompass
Change view
Click to view available options
Color:
Black
Catalog No. Color
19-048-692C Black
1 options
This item is not returnable. View return policy
Catalog No. 19-048-692C Supplier Valeo Supplier No. VA4575LG
This item is not returnable. View return policy
Gloves maximize breathability and comfort with an all mesh back and a soft terry lining.
  • Genuine leather with cotton mesh back
  • Soft terry lining for comfort and durability
  • Padded, leather palm
  • Hook & loop closure
Specifications
Back Material Mesh
Color Black
Hand Specific
Material Cowhide Leather
Product Type Fingerless Gloves
Size Large
Finger Style Fingerless
Glove Cuff Style Hook and Loop
Liner Material Soft Terry
Palm Type Padded
Series VI4862