Learn More
TARGETMOL CHEMICALS INC VALEPOTRIATE 5MG
SDP
Catalog No. 502579829
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Valepotriate (Valtrate) fraction can have sedative effects and affect behavioral parameters related to recognition memory. Valepotriates a new class of cytotoxic and antitumor agents they are very potent cytotoxic agents for the HTC hepatoma cells. Valepotriates may have a potential anxiolytic effect on the psychic symptoms of anxiety. purity: 99%
Catalog No. 50-257-9829 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S19995MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Valepotriate (Valtrate) fraction can have sedative effects and affect behavioral parameters related to recognition memory. Valepotriates a new class of cytotoxic and antitumor agents they are very potent cytotoxic agents for the HTC hepatoma cells. Valepotriates may have a potential anxiolytic effect on the psychic symptoms of anxiety. purity: 99%