Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Valeraldehyde

Catalog No. 1101321L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-013-21L 1 L
11-013-2250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-013-21L Supplier Sigma Aldrich Supplier No. 1101321L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Valeraldehyde

Specifications

CAS 110-62-3
Melting Point -92°C (lit.)
Density 0.81 g/mL (at 25°C (literature))
Boiling Point 102°C to 103°C (lit.)
Molecular Formula C5H10O
Refractive Index n20/D 1.394 (literature)
Linear Formula CH3(CH2)3 CHO
MDL Number MFCD00007026
Quantity 1 L
Synonym Pentanal
Molecular Weight (g/mol) 86.13
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.