Learn More
Sigma Aldrich Fine Chemicals Biosciences Valeraldehyde 97 fG1KG
Catalog No. 502897315
Encompass
Valeraldehyde is one of the key volatile flavor compounds identified in cooked rice beef fish meat and uncured ham.
Catalog No. 50-289-7315 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3098181KGK
Valeraldehyde is one of the key volatile flavor compounds identified in cooked rice beef fish meat and uncured ham.