Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valeraldehyde, 97%, Thermo Scientific™

Catalog No. AC149565000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC149565000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC149565000 Supplier Thermo Scientific Chemicals Supplier No. 149565000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-62-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007026
InChI Key HGBOYTHUEUWSSQ-UHFFFAOYSA-N
Synonym valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde
PubChem CID 8063
ChEBI CHEBI:84069
IUPAC Name pentanal
SMILES CCCCC=O

Specifications

Melting Point -92.0°C
Density 0.8070g/mL
Boiling Point 103.0°C
Flash Point 12°C
Refractive Index 1.3900 to 1.3950
Linear Formula CH3(CH2)3CHO
Quantity 500 mL
Specific Gravity 0.807
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: soluble in alcohol, ether, propylene glycol and, many organic solvents
Formula Weight 86.13
Chemical Name or Material Valeraldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.