Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valeramide | 626-97-1 | MFCD00041895 | ≥98.0% | 101.15 g/mol | C5H11NO | 25 G
SDP

Catalog No. 5000282880
Encompass_Preferred

Valeramide (pentanimidic acid) is a small-molecule raw material for organic synthesis, produced by ammonolysis of caproamide. It is used in the manufacture of polyamides, amide resins, preservatives, and nylon fibers, and is supplied as a white to off-white solid with high purity suitable for laboratory and industrial use.

  • CAS number 626-97-1.
  • Molecular formula C5H11NO.
  • Molecular weight 101.15 g/mol.
  • Purity ≥98.0%.
  • Appearance white to off-white solid.
  • Available package sizes include 5 g, 10 g, 25 g, 50 g, and 100 g.
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year; store dry at room temperature for up to 3 years.

Catalog No. 50-002-82880 Supplier Medchemexpress LLC Supplier No. HYW14403725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valeramide (pentanimidic acid) is a small-molecule raw material for organic synthesis, produced by ammonolysis of caproamide. It is used in the manufacture of polyamides, amide resins, preservatives, and nylon fibers, and is supplied as a white to off-white solid with high purity suitable for laboratory and industrial use.

  • CAS number 626-97-1.
  • Molecular formula C5H11NO.
  • Molecular weight 101.15 g/mol.
  • Purity ≥98.0%.
  • Appearance white to off-white solid.
  • Available package sizes include 5 g, 10 g, 25 g, 50 g, and 100 g.
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year; store dry at room temperature for up to 3 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.