Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valerate (sodium) | 6106-41-8 | 124.11 | 1 G
SDP

Catalog No. 5000460309
Encompass_Preferred

Valerate (sodium) is an ester product intended for research use only. This compound is a white to off-white solid with a purity of 98%. It is soluble in water at 200 mg/mL with sonication, and is stable under recommended storage conditions.

  • Purity: 98%
  • Appearance: White to off-white solid
  • Molecular formula: C5H9NaO2
  • Soluble in water (200 mg/mL with sonication)
  • Stable under recommended storage conditions
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year or -20°C for 6 months

Catalog No. 50-004-60309 Supplier Medchemexpress LLC Supplier No. HYW0070871G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valerate (sodium) is an ester product intended for research use only. This compound is a white to off-white solid with a purity of 98%. It is soluble in water at 200 mg/mL with sonication, and is stable under recommended storage conditions.

  • Purity: 98%
  • Appearance: White to off-white solid
  • Molecular formula: C5H9NaO2
  • Soluble in water (200 mg/mL with sonication)
  • Stable under recommended storage conditions
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year or -20°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.