Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valerate (sodium) | 6106-41-8 | 124.11 | 10 G
SDP

Catalog No. 5000460396
Encompass_Preferred

Valerate (sodium) is an ester product primarily used for research purposes.

  • Purity: 98%
  • Molecular weight: 124.11
  • Appearance: White to off-white solid
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year, -20°C for 6 months
  • Identified for use as laboratory chemical

Catalog No. 50-004-60396 Supplier Medchemexpress LLC Supplier No. HYW00708710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valerate (sodium) is an ester product primarily used for research purposes.

  • Purity: 98%
  • Molecular weight: 124.11
  • Appearance: White to off-white solid
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year, -20°C for 6 months
  • Identified for use as laboratory chemical

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.