Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valerate sodium | 6106-41-8 | 124.11 | 25 G
SDP

Catalog No. 5000448160
Encompass_Preferred

Valerate sodium is an ester product primarily intended for research use only, and not for patients. This compound has a molecular weight of 124.11 and a chemical formula of C5H9NaO2. It presents as a solid, ranging in color from white to off-white.

  • Purity: Valerate sodium has a reported purity of 98%.
  • Physical characteristics: It is a white to off-white solid.
  • Chemical identifiers: CAS no. 6106-41-8, molecular weight 124.11, formula C5H9NaO2.
  • Storage and handling: Store at room temperature, keep dry and cool. In solvent: -80°C for 1 year, or -20°C for 6 months.
  • Solubility: It is soluble in H2O at 200 mg/mL (1611.47 mM), requiring sonication.
  • Research use: Exclusively for research purposes; not for human use.

Catalog No. 50-004-48160 Supplier Medchemexpress LLC Supplier No. 25GHYW007087
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valerate sodium is an ester product primarily intended for research use only, and not for patients. This compound has a molecular weight of 124.11 and a chemical formula of C5H9NaO2. It presents as a solid, ranging in color from white to off-white.

  • Purity: Valerate sodium has a reported purity of 98%.
  • Physical characteristics: It is a white to off-white solid.
  • Chemical identifiers: CAS no. 6106-41-8, molecular weight 124.11, formula C5H9NaO2.
  • Storage and handling: Store at room temperature, keep dry and cool. In solvent: -80°C for 1 year, or -20°C for 6 months.
  • Solubility: It is soluble in H2O at 200 mg/mL (1611.47 mM), requiring sonication.
  • Research use: Exclusively for research purposes; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.