Learn More
Cayman Chemical Valerenic Acid, Purity: ≥98%, CAS: 3569-10-6, 10 mg
SDP
Catalog No. 502943409
Encompass_Preferred
A sesquiterpenoid with diverse biological activities; selectively RXRβ over RXRα and RXRγ in reporter assays using HEK293T cells (EC50s = 5.2, 27, and 43 µM, respectively), as well as RARs, PPARs, LXRs, FXR, CAR, VDR, and PXR at 30 µM; enhances GABA-induced activation of α1β2γ2-subunit containing GABA receptors in CHO cells (EC50 = 12.6 µM); cytotoxic to GLC-4 and COLO 320 cells (IC50s = 127 and 124 µM, respectively); decreases motor performance in the rotarod test and increases pentobarbital-induced sleeping time in mice at 100 mg/kg, i.p.; increases the number of entries into and percentage of time spent in the open arms of the elevated plus maze in mice, indicating anxiolytic-like activity, at 12 mg/kg
Catalog No. 50-294-3409 Supplier Cayman Chemical Supplier No. 4011310 MG
A sesquiterpenoid with diverse biological activities; selectively RXRβ over RXRα and RXRγ in reporter assays using HEK293T cells (EC50s = 5.2, 27, and 43 µM, respectively), as well as RARs, PPARs, LXRs, FXR, CAR, VDR, and PXR at 30 µM; enhances GABA-induced activation of α1β2γ2-subunit containing GABA receptors in CHO cells (EC50 = 12.6 µM); cytotoxic to GLC-4 and COLO 320 cells (IC50s = 127 and 124 µM, respectively); decreases motor performance in the rotarod test and increases pentobarbital-induced sleeping time in mice at 100 mg/kg, i.p.; increases the number of entries into and percentage of time spent in the open arms of the elevated plus maze in mice, indicating anxiolytic-like activity, at 12 mg/kg