Learn More
Sigma Aldrich Fine Chemicals Biosciences Valerenic acid phyproofR R10MG
Catalog No. 502902228
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-2228 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHL8928810MG
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG